Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Magnesiumstearat (pflanzliche Basis), Hyqual® (NF - GenAR®) NF, FCC, Macron Fine Chemicals™

Lieferant: MACRON AVANTOR BRAND

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


Veränderungsmanagement-Kategorie= R

2257-88EA 2497.64 EUR InStock
2257-88 2257-26 2257-06
Magnesiumstearat (pflanzliche Basis), Hyqual® (NF - GenAR®) NF, FCC, Macron Fine Chemicals™
Magnesiumstearat
Food GMP Manufactured Product
GMP Manufactured Product
Meets B.P. Chemical Specifications
Meets ChP Chemical Specifications
Meets E.P. Chemical Specifications
Meets F.C.C. Requirements
Meets J.P. Chemical Specifications
Meets N.F. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
Bulk Pharmaceutical Chemical


Formel: C₃₆H₇₀MgO₄
MW: 591.25 g/mol
Melting Pt: 130…140 °C
Dichte: 1.03 g/cm³ (20 °C)
Storage Temperature: Raumtemperatur
MDL Number: MFCD00036391
CAS-Nummer: 557-04-0
EINECS: 209-150-3
Merck Index: 13,05714
Order Now
Spezifikation Testergebnisse
Food GMP Manufactured Product
Meets N.F. Requirements
Meets E.P. Chemical Specifications
Meets J.P. Chemical Specifications 4.0 - 5.0 %
GMP Manufactured Product ≥ 40 %
Meets ChP Chemical Specifications ≥ 90 %
Meets B.P. Chemical Specifications Passes Test
Meets F.C.C. Requirements Passes Test
CAUTION: For Manufacturing, processing or repackaging Passes Test
Bulk Pharmaceutical Chemical Passes Test
Bulk Food Chemical ≤ 1000
NF - Assay (as as Mg) (dried basis) 4.0 - 5.0 %
NF - Stearic Acid ≥ 40 %
NF - Stearic and Palmitic Acids ≥ 90 %
NF - Identification A Passes Test
NF - Identification B Passes Test
NF - Acidity or Alkalinity Passes Test
NF - Microbiological Passes Test
NF - Total Aerobic Microbial Count (cfu/g) ≤ 1000
NF - Total Mold and Yeast Count (cfu/g) ≤ 500
NF - Salmonella Species (negative) Passes Test
NF - Escherichia Coli (negative) Passes Test
NF - Loss on Drying at 105°C ≤ 6.0 %
Specific Surface Area, m²/g 5.0 - 10.0
NF - Limit of Chloride ≤ 0.1 %
NF - Limit of Sulfate ≤ 1.0 %
NF - Cadmium (Cd) ≤ 3 ppm
NF - Lead (Pb) ≤ 10 ppm
NF - Nickel (Ni) ≤ 5 ppm
EP/BP - Identification C Passes Test
EP/BP - Identification D Passes Test
EP/BP - Acidity or Alkalinity Passes Test
EP/ BP - Chloride (Cl) ≤ 0.1 %
EP/BP - Sulfate (SO₄) ≤ 1.0 %
EP/BP - Cadmium (Cd) ≤ 3 ppm
EP/BP - Lead (Pb) ≤ 10 ppm
EP/BP - Nickel (Ni) ≤ 5 ppm
EP/BP - Loss on Drying at 105°C ≤ 6.0 %
Total Aerobic Microbial Count (cfu/g) ≤ 1000
Escherichia Coli (negative) Passes Test
EP/BP - Assay (as as Mg) (dried basis) 4.0 - 5.0 %
Salmonella (absent) Passes Test
EP/BP - Stearic Acid ≥ 40.0 %
EP/BP - Stearic and Palmitic Acids ≥ 90.0 %
Total Mold and Yeast Count (cfu/g) ≤ 100
JP - Identification Passes Test
JP - Acidity or Alkalinity Passes Test
JP - Chloride (Cl) ≤ 0.1 %
JP - Sulfate (SO₄) ≤ 1.0 %
JP - Heavy Metals (as Pb) ≤ 20 ppm
JP - Stearic Acid ≥ 40 %
JP - Stearic and Palmitic Acids ≥ 90 %
JP - Loss on Drying at 105°C ≤ 6.0 %
JP - Microbiological Passes Test
JP - Total Aerobic Microbial Count (cfu/g) ≤ 1000
JP - Total Mold and Yeast Count (cfu/g) ≤ 500
JP - Salmonella Species (negative) Passes Test
JP - Escherichia Coli (negative) Passes Test
JP - Assay (as as Mg) (dried basis) 4.0 - 5.0 %
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Assay (as MgO) 6.8 - 8.3 %
FCC - Lead (Pb) ≤ 5 mg/kg
FCC - Loss on Drying at 105°C ≤ 4.0 %
Thru U.S. No. 325 Sieve, For Information Only ≤ 0.10 %
Tapped Density (g/cc) 0.21 - 0.33
Apparent (Bulk) Density 0.07 - 0.17 g/cm3
Particle Size (50th percentile) (µm) 10.5 - 16.5
Particle Size (90th percentile) (µm) ≤ 35
ChP - Identification 1 Passes Test
ChP - Identification 2 Passes Test
ChP - Acidity or Alkalinity Passes Test
ChP - Chlorides (Cl) ≤ 0.10 %
ChP - Sulfates (SO₄) ≤ 0.6 %
ChP - Loss on Drying @ 80° C ≤ 5.0 %
ChP - Iron (Fe) ≤ 0.01 %
ChP - Cadmium (Cd) ≤ 0.0003 %
ChP - Nickel (Ni) ≤ 0.0005 %
ChP - Heavy metals ≤ 0.001 %
ChP - Stearic Acid ≥ 40.0 %
ChP - Stearic and Palmitic Acids ≥ 90.0 %
ChP - Total Aerobic Microbial Count ≤ 1000 cfu/g
ChP - Total Yeast and Mold ≤ 100 cfu/g
ChP - Salmonella None Detected
ChP - Escherichia Coli None Detected
ChP - Assay (as Mg) (dried basis) 4.0 - 5.0 %
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.

 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge