Tubing holder for beakers, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
F182350000
229-0001EA 79.1 EUR InStock
229-0001
Tubing holder for beakers, SP Bel-Art
Klemmen Schlauchklemmen und Sanitärklemmen
This PP clip holds flexible tubing steady and fits securely onto the edge of beakers or pitchers up to ¼" thick.
  • 5,1 cm (2") long arm has a hole to accept tubing up to 9,5 mm (⅜") Ext. Ø
  • Autoclavable
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge