Dialysis bag clips, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
F182370000
229-0003EA 61.4 EUR InStock
229-0003
Dialysis bag clips, SP Bel-Art
Klemmen Klemmen für den allgemeinen Gebrauch
These strong clips made from PP are ideal for suspending dialysis bags in large beakers or pitchers up to 0,63 cm (¼") thick.
  • Bags slide easily into the slot and then the holder can be clipped onto the container’s edge
  • Smooth PP surface may be marked with identifying sequence numbers
  • Autoclavable
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge