Tubing clips
Lieferant: MURRAY S
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
L145/01
229-0589EA 50.5 EUR InStock
229-0589
Tubing clips
Klemmen Schlauchklemmen und Sanitärklemmen
Acetal copolymer.
  • For rubber and flexible PVC tubing with an external diameter of up to 14 mm
  • Hinged side allows it to be clamped around the tubing already connected to the apparatus
  • Screw adjustment ensures fine flow control
  • Material is easy to clean, sterilisable, and resistant to most acids and alkalis
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge