Schlauchverbinder, X-Form
Lieferant: KARTELL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
71410 71411 71412 71413 71414 71415 71416
229-0742EA 27.3 EUR InStock
229-0742 229-0743 229-0744 229-0745 229-0746 229-0747 229-0748
Schlauchverbinder, X-Form
Fittings und Verbinder
PP, durchscheinend
  • Good mechanical strength and chemical resistance
  • Very good thermal resistance
  • Autoclavable
  • Suitable for foodstuff
Dimensions are shown as both nominal and exact as elasticity of tubing will influence the choice of size. Use our tubing selector 229-0111 to determine "grip". This will then give equivalent crest dimension for the particular connector concerned.
They meet with the Food and Drug regulations.90/128CEE 92/39CEE and FDA 21 CFR 177.1520.
Suitable for foodstuff.
Zertifizierungen: Erfüllen die Normen gemäß FDA.90/128 CEE, 92/39 CEE und FDA 21 CFR 177.1520.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge