Tubing connectors, T-shaped
Lieferant: PIEPER FILTER
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
PP-S16S16T PP-S24S24T PP-S32S32T PP-S40S40T PP-S48S48T1
229-4131EA 45.7 EUR InStock
229-4131 229-4132 229-4133 229-4134 229-4135
Tubing connectors, T-shaped
Fittings und Verbinder
Mini connectors for tubing, made of PP, translucent, T-shape.
  • Good temperature resistance
  • Good chemical resistance
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge