22922.K2
22922.M1
22922.K2EA
86.2
EUR
InStock
22922.K2
22922.M1
Chlorbenzol ≥99,5% für die HPLC
Chlorbenzol
Purity: 99.5%
CAS No.: 108-90-7
Molecular Formula: C6H5Cl
Formula Weight: 112.56
Flash Point: 28°(82°F)
Physical Form: Liquid
Density: 1.106
Refractive Index: 1.5240
Appearance: Colourless
Odour: Aromatic
Melting Point: -45°
Boiling Point: 132°
Beilstein Reference: 605632
Merck Reference: 14,2121
UN No.: 1134
MDL No.: MFCD00000530