Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Indigo 95% (Farbstoffgehalt), Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Achtung

M-Clarity™ Qualitätsstufe = MQ200

229296-100G 229296-25G
229296-100GEA 142 EUR InStock
229296-100G 229296-25G
Indigo 95% (Farbstoffgehalt), Sigma-Aldrich®
Indigo
Indigo has been used as a dye to measure food consumption in drosophila. It has also been used to investigate the ability of fungal strains in the degradation and decolorisation of indigo dye.

Indigo is a rich blue color dye. It can be extracted from plants of genus indigofera and can also be produced synthetically. Indigo is a vat textile dye, widely used on cellulosic fibers like cotton. Additionally, it is also used to inhibit corrosion of mild steel in aerated sulfuric acid.


Formel: C₁₆H₁₀N₂O₂
Melting Pt: >300 °C (Lit.)
Dichte: 1,35 g/cm³ (20 °C)
Storage Temperature: Room Temperature
MDL Number: MFCD00005722
CAS-Nummer: 482-89-3
EINECS: 207-586-9
Merck Index: 12,04977
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e4c344ac5f2c955a7b20d49372b316ace2bc416&vid=1e4c344ac5f2c955a7b20d49372b316ace2bc416&prid=web&clid=SAPDAM
Order Now
Spezifikation Testergebnisse
Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Wavelength in chloroform 599 - 605 nm
Extinction Coefficient ≥13500
Wavelength 282 - 288 nm
Carbon 68.9 - 77.7%
Nitrogen 10.0 - 11.3%

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge