22967.18
22967.30
22967.18EA
90.4
EUR
InStock
22967.18
22967.30
Tetraethylorthosilikat (TEOS) ≥99,999% (Metall-Basis)
Tetraethylorthosilikat (TEOS)
Purity: 99.999+%
CAS No.: 78-10-4
Molecular Formula: C8H20O4Si
Formula Weight: 208.33
Flash Point: 46°(116°F)
Physical Form: Liquid
Density: 0.934
Refractive Index: 1.3820
Sensitivity: Moisture Sensitive
Appearance: Colourless
Odour: Pleasant
Melting Point: -85°
Boiling Point: 168°
Beilstein Reference: 1422225
Merck Reference: 14,3851
UN No.: 1292
MDL No.: MFCD00009062