Sampling scoops with lid, Sterileware®, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
H369220000 H369240000
231-0636EA 681 EUR InStock
231-0636 231-0637
Sampling scoops with lid, Sterileware®, SP Bel-Art
Schaufeln
White PS. These scoops made of high-impact FDA grade PS feature a detachable lid. They eliminate the cost of separate sterile bags or containers, saving you time and money in your sample collection process.
  • Longer handle increases the distance between the hand and sample to reduce contamination potential
  • Scoops remain upright when placed on a flat surface; can be used as weighing boats
  • Lock-tab on the lid can be bent under for a tighter seal
  • Sampling data can be marked on the lid with wax pencil, pre-marked label or labeling tape to facilitate tracking
  • Individually sealed and sterilised in a polyethylene bag ready for one time use and disposal
  • Lot stamped for accurate lot tracing; shipped with a Certificate of Processing for Sterilization
To use, simply remove the scoop from the bag and detach the lid using the convenient tab. After taking the sample, replace the snap-on lid and use the scoop as a container for transport.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge