Powder funnels, PP, SP Bel-Art Sterileware®
Lieferant: Bel-Art Products, a Part of SP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
F14660-1065 F14660-1080 F14660-1100 F14660-1150
231-0648EA 745 EUR InStock
231-0648 231-0649 231-0650 231-0651
Powder funnels, PP, SP Bel-Art Sterileware®
Trichter Pulvertrichter
These sterile, wide stem funnels are ideal for transferring powders or granules using a sterile sampling tool into a sterile container.
  • 60° angle and large bore stem help minimise bridging of powder
  • FDA grade polypropylene; safe for food, drug and cosmetic use
  • Gamma irradiated sterile (SAL 10⁻⁶)
  • Individually bagged; ready for one time use and disposal
Lot stamped for accurate tracing; shipped with a Certificate of Processing for Sterilisation.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge