23111.18
23111.30
23111.18EA
100
EUR
InStock
23111.18
23111.30
Cobalt(II) phosphate octahydrate
Cobalt(II) phosphate octahydrate
CAS No.: 10294-50-5
Molecular Formula: Co3(PO4)2.8H2O
Formula Weight: 510.87 (366.74anhy)
Physical Form: Powder
Density: 2.769
Odour: Odourless
MDL No.: MFCD00016028