23141.14
23141.22
AB204251.0025
23141.14EA
102
EUR
InStock
23141.14
23141.22
ABCRAB204251.0025
Cobalt(II)oxid ≥95%
Cobalt(II)oxid
Purity: 95%
CAS No.: 1307-96-6
Molecular Formula: CoO
Formula Weight: 74.93
Physical Form: Powder
Density: 6.45
Appearance: Black-green
Melting Point: 1935°
Solubility: Soluble in acids or alkalies. Insoluble in water
Merck Reference: 14,2446
UN No.: 2811
MDL No.: MFCD00016031
Pigments, glass Colouring, semiconductors