Surgical scalpels and blades, Sabre
Lieferant: SWANN MORTON
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
0570 0565 0563
SWAN0570EA 18 EUR InStock
SWAN0570 SWAN0565 233-0163
Surgical scalpels and blades, Sabre
Skalpelle Skalpelle, komplett
Carbon steel blades. These blades are available separately without handles or as ready-to-use scalpels with PS handle. They are ideal for use during Podiatry and Chiropody treatments.
  • Scalpels are sterile and diposable
  • PS handles carry a graduated measure (for indication only) on the reverse side
  • Closed edge blades are ideal for 'paring' or 'scraping' skin within Podiatry and Chiropody treatments
Verpackung: Sterile scalpels are individually packaged into a transparent thermo formed tray providing improved product visibility.
Blades only are available in individual foil packs (sterile) or in foil packets of five (non sterile).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge