Blades for safety knives, No. 142
Lieferant: MARTOR ARGENTAX
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
142.70
233-2238EA 17.5 EUR InStock
233-2238
Blades for safety knives, No. 142
Rasiermesser und Klingen
Non corrosive trapezoid blade for use with safety knives.
  • Deep-edged, two-sided ground-edge
  • Double bevel edge-ground
  • Two-times usable blade
Mountable in:
SECUNORM PROFI
SECUNORM IDEAL
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge