Economy water vapor trap
Lieferant: Sigma-Aldrich
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
23988
23988.EA 215 EUR InStock
23988.
Economy water vapor trap
Filter Gasfilter
This economy water vapor trap is designed for moisture removal for compressed air service to pneumatically-controlled devices.
  • Purifier body made of clear polycarbonate
  • Trap contains a mixture of Molecular Sieves 4A and 13X
  • Not to be used with carrier gases due to permeability of purifier body
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge