Tee connectors, Stronghold™
Lieferant: HARVARD APPARATUS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
53-2032-W
241-2241EA 22.4 EUR InStock
241-2241
Tee connectors, Stronghold™
Stativstäbe
The main use of this tee connector is to mount clamps and adapters that have a male connector to vertical lattice rod. One end has an open side that will clamp on a rod up to 13 mm Ø. The other end has a female socket 19 mm deep that will take a male connector, or the butt end of a rod, up to 13 mm.
  • Precision injection molded nylon with 30% glass fiber reinforcement provides enormous strength at a great saving in weight
  • There is positively no corrosion possible
  • Material is perfect for laboratory use since it has the very high melting point of 221 °C and good resistance to chemicals, particularly salt solutions and most solvents
  • Colours are an integral part of the product, there is no paint, plating or coating that can fail in use
  • Available with red body and white thumb screws and with white body and red thumb screws
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge