Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Micro air sampler, PAS-500, Supelco®

Lieferant: Sigma-Aldrich

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

24865
24865.EA 1250 EUR InStock
24865.
Micro air sampler, PAS-500, Supelco®
Probenehmer Luftkeimsammler
This low flow pump is lightweight (4 oz.) and compact (7" H) with a flame-retardant case, fitting easily into your shirt pocket. The adsorbent tube connects directly to the inlet of the pump.

  • The unit is powered by a convenient
  • Easily replaceable 9 V battery

A full flow regulation feature provides constant voltage to the pump, even as the battery voltage drops. This sampler is intrinsically safe but not suitable for sampling in explosive environments (NOT EX-Rated). The design features a current-limiting resistor in the battery circuit which prevents danger of short-circuit spark.

The PAS-500 sampler is versatile and adapts to fit both 6 and 8 mm diameter tubes. The flow range is 5 to 200 cc/min. An optional low flow adapter enables you to sample at 20 cc/min.

Zertifizierungen: CE approved.

Lieferumfang: Includes 6 mm tube holder, screwdriver and two 9-volt batteries.

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge