MEM amino acids 50X solution without L-glutamine, Corning®
Lieferant: MEDIATECH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
25-030-CI
25-030-CIEA 138.9 EUR InStock
25-030-CI
MEM amino acids 50X solution without L-glutamine, Corning®
Zellkultur-Ergänzungen und -Zusätze
The addition of MEM nonessential amino acids to medium can improve growth and the viability of propagating cells in culture while reducing the biosynthetic burden on these cells.
This concentrated supplement allows for efficient addition to media with no risk of dilution of media components.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge