Corning® Trypsin
Lieferant: MEDIATECH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
25-050-CI
25-050-CIEA 78.11 EUR InStock
25-050-CI
Corning® Trypsin
Enzyme
0,25% Trypsin in HBSS (Hank’s Balanced Salt Solution) without calcium and magnesium.
  • Porcine parvovirus tested
  • 6×100 ml
Trypsin is an enzymatic reagent that is used to release adherent cells from tissue plates for passaging. This offering contains trypsin at a concentration of 0,25%.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge