Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Wasser, Zellkultur-Qualität, getestet nach USP Vorgaben zur Injektion, steril for sterile Water for Injection , Corning®

Lieferant: MEDIATECH

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


25-055-LH 25-055-LG 25-055-LB 25-055-CVC 25-055-CM 25-055-CI 25-055-CV
MDTC25-055-LHEA 1350 EUR InStock
MDTC25-055-LH MDTC25-055-LG MDTC25-055-LB MDTC25-055-CVC 25-055-CM MDTC25-055-CI 25-055-CV
Wasser, Zellkultur-Qualität, getestet nach USP Vorgaben zur Injektion, steril for sterile Water for Injection , Corning®
Wasser
Corning® Cell Culture Grade Water Tested to USP Sterile Water for Injection Specifications. Cell culture grade water is high-quality water suitable for use as a solvent in the preparation of cell culture media and other reagents for sensitive applications.

  • Complete Certificate of Analysis available for each production lot
  • Produced under the highest industry standards to ensure superior results
  • Meets WFI Standards

Our bioprocess container selections are suitable for large-scale applications such as cleaning and upstream and downstream reagent preparation. Cell culture grade water goes through a wide array of testing which includes testing to the chemical and physical properties found in the USP and EP monographs for sterile Water for Injection (WFI).


Formel: H₂O
MW: 18.02 g/mol
Siedepunkt: 100
Melting Pt: 0
Storage Temperature: Raumtemperatur
MDL Number: MFCD00011332
CAS-Nummer: 7732-18-5
Merck Index: 13,10098
Order Now
Spezifikation Testergebnisse
Mycoplasma Tested negative
Plating efficiency Pass
Oxidizable substances Solution remains faintly pink
Total organic carbon (before packaging) USP <643> < 500 ppb
Water conductivity (packaged water) USP <645> Meets USP Requirements
Water conductivity (before packaging) USP <645> Meets USP temperature/conductivity requirements
Endotoxin USP <85> ≤ 0.025 EU/mL
Sterility USP <71> Pass
Particulate matter USP <788> ≤ 25 particles/mL that are ≥ 10 μm in size
≤ 3 particles/mL that are ≥ 25 μm in size

 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge