Cell dissociation reagent, Corning® Cellstripper®
Lieferant: MEDIATECH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
25-056-CI
25-056-CIEA 113.08 EUR InStock
25-056-CI
Cell dissociation reagent, Corning® Cellstripper®
Zellkultur-Ergänzungen und -Zusätze
Cellstripper® is a non-enzymatic cell dissociation solution formulated with a proprietary mixture of chelators that gently dislodge adherent cells in tissue culture. It is an alternative to trypsin when used with either serum-free or serum-supplemented media. Because Cellstripper® is a gentler reagent than trypsin, cells can be exposed to Cellstripper for longer periods of time. Cellstripper® achieves superior results in comparison to enzymatic dissociation products.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge