Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Wasser, erfüllt analytische Spezifikationen von Ph. Eur., USP zur Injizierung (WFI), Reagenz für die Zellkultur, steril, Corning®

Lieferant: MEDIATECH

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


25-065-LB 25-065-LG 25-065-LH
25-065-LBEA 493.16 EUR InStock
25-065-LB 25-065-LG 25-065-LH
Wasser, erfüllt analytische Spezifikationen von Ph. Eur., USP zur Injizierung (WFI), Reagenz für die Zellkultur, steril, Corning®
Wasser
Cell culture grade water is high-quality water suitable for use as a solvent in the preparation of cell culture media and other reagents for sensitive applications. Our bioprocess container selections are suitable for large-scale applications such as cleaning and upstream and downstream reagent preparation. Cell culture grade water goes through a wide array of testing which includes testing to the chemical and physical properties found in the USP and EP monographs for sterile Water for Injection (WFI).

Technical Advantage: Wide range of applications in processes requiring very high levels of sterility USP/EP Certified Water is validated to the most recent specifications found in the United States Pharmacopeia (USP) and the European Pharmacopeia (EP). The specific testing is in accordance to testing for "Sterilized Water for Injection", (European Pharmacopeia), and "Sterile Water for Injection", (United States Pharmacopeia).


Formel: H₂O
MW: 18.02 g/mol
Siedepunkt: 100
Melting Pt: 0
Storage Temperature: Raumtemperatur
MDL Number: MFCD00011332
CAS-Nummer: 7732-18-5
Merck Index: 13,10098
Order Now
Spezifikation Testergebnisse
Mycoplasma Tested Negative
LM (plating effciency) Pass
Oxidizable Substances Pass
Total Organic Carbon (before packaging) USP <500 ppb
Water Conductivity (packaged water) USP Meets USP requirements
Water Conductivity (before packaging) USP Meets USP Temperature/Conductivity Requirements
Endotoxin USP ≤0.025 EU/ml
Sterility USP Pass
Acidity-Alkalinity Tested in Accordance with EP
Chlorides Pass
Nitrates ≤0.2 ppm
Sulphates Pass
Ammonium ≤0.2 ppm
Calcium and Magnesium Pass
Residue on Evaporation ≤3 mg (0.003)

 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge