M-Clarity™ Qualitätsstufe = MQ100
252751-25G
252751-5G
252751-25GEA
245
EUR
InStock
252751-25G
252751-5G
3-Ethoxy-4-methoxybenzaldehyd ≥99%, Sigma-Aldrich®
3-Ethoxy-4-methoxybenzaldehyde
3-Ethoxy-4-methoxybenzaldehyde is a derivative of vanillin. 3-Ethoxy-4-methoxybenzaldehyde may be used to synthesise Compound of Designation red 10 binding (CDr10b) to investigate its role in neuroinflammatory diseases.