Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

D(-)-Mannitol 97,0-102,0% (Trockenmasse), Pulver USP, Multikompendial GMP Manufactured Product, J.T.Baker®

Lieferant: AVANTOR PERFORMANCE MATERIALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


Veränderungsmanagement-Kategorie= R

2553-01EA 253.04 EUR InStock
2553-01 2553-05 2553-07 2553-09 2553-32 2553-34 2553-37 2553-39 2553-38
D(-)-Mannitol 97,0-102,0% (Trockenmasse), Pulver USP, Multikompendial GMP Manufactured Product, J.T.Baker®
D(-)-Mannitol


Formel: HOH₂C(CH(OH))₄CH₂OH
MW: 182.17 g/mol
Siedepunkt: 290…295 °C (4 hPa)
Melting Pt: 164…169 °C
Dichte: 1.52 g/cm³
Storage Temperature: Raumtemperatur
MDL Number: MFCD00064287
CAS-Nummer: 69-65-8
EINECS: 200-711-8
Merck Index: 13,05769
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=163f556dca45086e05ff490ddc8eeb65d04aea05&vid=a880987b6e666d8d6d0f15d1877728a17c3a02ad&prid=web&clid=SAPDAM
Order Now
Spezifikation Testergebnisse
GMP Manufactured Product 97.0 - 102.0%
Meets E.P. Chemical Specifications Passes Test
Meets U.S.P Requirements ≤2.0%
Meets B.P. Chemical Specifications ≤2.0%
Meets J.P. Chemical Specifications ≤0.10%
CAUTION: For Manufacturing, processing or repackaging ≤2.0%
Bulk Pharmaceutical Chemical ≤0.5%
USP - Assay (dried basis) 97.0 - 102.0 %
USP - Identification Passes Test
USP - Related Substances: Sorbitol ≤ 2.0 %
USP - Related Substances: Sum of isomalt& maltitol ≤ 2.0 %
USP - Related Substances: Unspecified impurities ≤ 0.10 %
USP - Related Substances: Total impurities ≤ 2.0 %
USP - Loss on Drying at 105°C ≤ 0.5 %
USP - Melting Point 165 - 170 °C
USP - Reducing Sugars ≤ 0.1 %
USP - Nickel (Ni) ≤ 1 ppm
USP - Appearance of Solution Passes Test
USP - Conductivity (uS cm-1) ≤ 20 µS/cm
USP - Total Aerobic Microbial Count ≤ 100 cfu/g
USP - Total Yeast and Mold Count ≤ 100 cfu/g
USP - Escherichia coli Passes Test
USP - Bacterial Endotoxins, IU/g ≤ 2.5
EP/BP - Assay (as HOCH₂(CHOH)₄CH₂OH) (dried basis) 97.0 - 102.0 %
EP/BP - Identification C Passes Test
EP/BP - Melting Point 165 - 170 °C
EP/BP - Appearance of Solution Passes Test
EP/BP - Conductivity, uS cm-1 ≤ 20
EP/BP - Reducing Sugars ≤ 0.1 %
EP/BP - Related Substances: Impurity A ≤ 2.0 %
EP/BP - Related Substances: Sum of Impurities B & C ≤ 2.0 %
EP/BP - Related Substances: Unspecified Impurities, each ≤ 0.10 %
EP/BP - Related Substances: Total Impurities ≤ 2.0 %
EP/BP - Loss on Drying ≤ 0.5 %
EP/BP - Escherichia coli None Detected
EP/BP - Salmonella None Detected
EP/BP - Total Yeast and Mold Count ≤ 100 cfu/g
EP/BP - Total Aerobic Microbial Count ≤ 100 cfu/g
EP/BP Endotoxin Concentration, <2.5 IU/g Passes Test
JP - Assay (dried basis) 97.0 - 102.0 %
JP - Identification Passes Test
JP - Related Substances: Sorbitol ≤ 2.0 %
JP - Related Substances: Sum of isomalt & maltitol ≤ 2.0 %
JP - Related Substances: Unspecified impurities ≤ 0.10 %
JP - Related Substances: Total impurities ≤ 2.0 %
JP - Heavy Metals (as Pb) ≤ 5 ppm
JP - Loss on Drying at 105°C ≤ 0.5 %
JP - Melting Point 165 - 170 °C
JP - Glucose ≤ 0.1 %
JP - Nickel (Ni) ≤ 1 ppm
JP - Clarity and Color of Solution Passes Test
JP - Conductivity (uS cm-1) ≤ 20 µS/cm
Suitable for use in the manufacture of parenteral dosage forms.
Maximum Endotoxin Concentration: 2.5 IU/g
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental impurities for this product is available on the associated Product Regulatory Data Sheet and elemental impurity profile report.

 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge