Deaza dGTP
Lieferant: GE HEALTHCARE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
27-2090-02
27-2090-02EA 337 EUR InStock
27-2090-02
Deaza dGTP
Nukleinsäure-Reagenzien Nukleotide
Band compressions are ambiguous band patterns on sequencing gels caused by stable intra-strand secondary structures that are not fully denatured during electrophoresis. Their occurrence can be reduced by substituting nucleotide analogues in the sequencing mixes. 7-Deaza dGTP (c⁷dGTP) can be used to resolve G-band compressions.
  • For reducing “band compressions” in sequencing gels
  • Supplied as a 5 mM solution
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge