DNA (Deoxyribonucleic acid) (aus Kalbsthymus), aktiviert
Lieferant: GE HEALTHCARE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
27-4575-01
27-4575-01EA 366 EUR InStock
27-4575-01
DNA (Deoxyribonucleic acid) (aus Kalbsthymus), aktiviert
DNS (Desoxyribonukleinsäure)
Activated calf thymus DNA at 25 A₂₆₀ units/ml (treated briefly with DNase I) for preparing substrate used in the study of DNA polymerases.
  • Nonmethylated λ DNA (at a concentration of 500 µg/ml) for preparation of molecular weight markers
  • High-quality DNAs from viral and eukaryotic sources used for preparation of assay reagents
Methylated DNA and nonmethylated DNA (at a concentration of 500 µg/ml) for determining methylation sensitivity of restriction enzymes.
MDL Number: MFCD00130922
CAS-Nummer: 9007-49-2
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge