Clamps for conical standard ground joints, KECK™, DURAN®
Lieferant: DWK Life Sciences
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
286336109 286336203 286336306 286336409 286336503 286336606 286336709 286336803 286336906
271-7111EA 3.7 EUR InStock
271-7111 271-7112 271-7113 271-7114 271-7115 271-7116 271-7107 271-7108 271-7117
Clamps for conical standard ground joints, KECK™, DURAN®
Klemmen Schliffklemmen
POM. These clips made of temperature resistant polyacetal (POM) are easy, safe and can be fitted quickly.
  • Good spring properties
  • Colour coded by size
  • No risk of scratching the glass
  • Will not corrode or bend out of shape
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge