Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Plasmid purification media, PlasmidSelect Xtra

Lieferant: GE HEALTHCARE

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


28-4024-02
28-4024-02EA 824 EUR InStock
28-4024-02
Plasmid purification media, PlasmidSelect Xtra
Protein-Aufbereitungs Protein Purification Resins
PlasmidSelect Xtra is a thiophilic aromatic adsorption chromatography medium with a selectivity that allows generic purification of supercoiled covalently closed circular forms of plasmid DNA to be separated from open circular forms.

  • Allows for generic purification of supercoiled plasmid DNA
  • Allows for consistent purification results when scaling up from research scale to cGMP production
  • Hydrophilic nature of the base matrix ensures low levels of non specific binding
  • BioProcess medium supported for industrial applications

Available in convenient prepacked format for research, process development and quantitative and qualitative analysis of supercoiled plasmid DNA.

Degree of substitution: Approx. 3,5 mg 2-mercaptopyridine/ml medium
Pressure/flow specification: <120 cm/h, XK 16/20 column, bed height 15 cm
Storage conditions: 4 to 30 °C, 20% ethanol
Chemical stability: stable in commonly used aqueous buffers


Order Now

 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge