Affinity chromatography columns, HiTrap™ AVB Sepharose™ HP
Lieferant: Cytiva
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
28-4112-11
28-4112-11EA 1176 EUR InStock
28-4112-11
Affinity chromatography columns, HiTrap™ AVB Sepharose™ HP
Chromatographiesäulen
AVB Sepharose High Performance is an affinity resin designed for the purification of adeno associated virus (AAV), Prepacked HiTrap columns for simple operation with a syringe, pump, or chromatographic system.
  • One step purification of adeno associated viruses
  • Affinity resin with high selectivity and low background binding
  • Large surface area to give enhanced capacity
AVB Sepharose High performance is an affinity resin with affinity for adeno associated viruses. It enables purification of adeno associated viruses in one step with high purity and excellent yields. It has proven affinity for adeno associated viruses from subclasses 1, 2, 3, and 5, but it is expected to have affinity for most of the other adeno associated virus subclasses. The ligand is a 14 kD fragment from a single chain antibody, expressed in yeast.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge