Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-IgG Goat Polyclonal Antibody (Cy3®)

Lieferant: Cytiva

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

28-9011-06
28-9011-06EA 483 EUR InStock
28-9011-06
Anti-IgG Goat Polyclonal Antibody (Cy3®)
Antikörper
ECL Plex Cy-conjugated Antibodies are part of an optimized system for quantitative analysis using fluorescent western blotting.

Optimized system for quantitative analysis using fluorescent Western blotting. ECL Plex Western Blotting System is optimized for single protein detection as well as multiplex protein detection using ECL Plex CyDye-Conjugated Secondary Antibodies and Hybond-LFP (low-fluorescent PVDF membrane) or Hybond ECL (nitrocellulose membrane). The optimized antibodies show high sensitivity, a very good dynamic range and low or no cross-reactivity. CyDye technology is dependable, reliable, and ensures accurate quantitation. The system is optimized on the Typhoon 9410 Scanner but is compatible with many fluorescent laser scanners and CCD cameras.

ECL Plex Western Blotting System is optimized for single protein detection as well as multiplex protein detection using ECL Plex CyDye-Conjugated Secondary Antibodies and Hybond-LFP (low-fluorescent PVDF membrane) or Hybond ECL (nitrocellulose membrane). The optimized antibodies show high sensitivity, a very good dynamic range and low or no cross-reactivity. CyDye technology is dependable, reliable, and ensures accurate quantitation. The system is optimized on the Typhoon 9410 Scanner but is compatible with many fluorescent laser scanners and CCD cameras.

Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Goat
Isotype: none
Reactivity: Rabbit

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge