illustra™ ProbeQuant™ G-50 micro columns
Lieferant: GE HEALTHCARE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
28-9034-08
28-9034-08EA 267 EUR InStock
28-9034-08
illustra™ ProbeQuant™ G-50 micro columns
Nukleinsäure-Reagenzien Reinigungskits und Reagenzien für Nukleinsäure
ProbeQuant™ G-50 micro columns use spin column chromatography to provide a fast and reliable method for purifying labelled probes from unincorporated labelled nucleotides. In addition, a portion of the eluted DNA can be counted in a scintillation counter and compared to an unpurified sample to give an approximation of percent incorporation. The columns are designed for use in a microcentrifuge, come prepacked with Sephadex™ G-50 DNA grade and are pre-equilibrated in STE, which means that they are ready to use. They can accommodate samples ranging in volume from 25 μl to 50 μl.
  • For rapid purification of labeled DNA (20 bases) from unincorporated labeled nucleotides in a volume of 25 to 50 μl using spin-column chromatography
  • Prepacked with Sephadex G-50 DNA Grade (see illustra Sephadex G-50 and G-100 DNA Grade) and pre-equilibrated in STE containing 0.15% Kathon CG/ICP Biocide
  • Requires less than 4 minutes from sample application to collection of purified product
  • Designed for use in a microcentrifuge
  • Tested in nickase, single and double-stranded exonuclease and RNase assays
In addition, a portion of the eluted DNA can be counted in a scintillation counter and compared to an unpurified sample to give an approximation of percent incorporation. The columns are designed for use in a microcentrifuge, come prepacked with Sephadex G-50 DNA grade and are pre-equilibrated in STE, which means that they are ready to use. They can accommodate samples ranging in volume from 25 to 50 μl.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge