Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Affinity chromatography column, HiScreen™ MabSelect Xtra™

Lieferant: Cytiva

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

28-9269-76
28-9269-76EA 1037 EUR InStock
28-9269-76
Affinity chromatography column, HiScreen™ MabSelect Xtra™
Chromatographiesäulen
HiScreen™ MabSelect Xtra™ is prepacked with MabSelect Xtra™ of 4,7 ml volume and bed height of 10 cm make HiScreen™ columns excellent tools for method optimisation, parameter screening for capture of monoclonal antibodies (MAbs), robustness testing, and convenient scale-up.

  • Easily connected in series to achieve 20 cm bed height
  • Small bed volume gives fast results and minimal sample/buffer consumption
  • Reproducible results, scalable to BioProcess columns packed with the same chromatography media using the same linear fluid velocity
  • Flow velocity <300 cm/h; the pressure over the packed bed varies depending on a range of parameters such as the characteristics of the chromatography medium and the column tubing used
  • Dynamic capacity: ~40 mg human IgG/ml medium

Process fluid velocities can be applied, since the 10 cm bed height gives enough residence time and the results can then serve as basis for linear process scale-up.

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge