Cation exchange chromatography column, HiPrep™ IEX SP Sepharose™ XL
Lieferant: Cytiva
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
28-9365-40
28-9365-40EA 745 EUR InStock
28-9365-40
Cation exchange chromatography column, HiPrep™ IEX SP Sepharose™ XL
Chromatographiesäulen
HiPrep™ SP XL 16/10 is a prepacked, ready to use column for ion exchange chromatography.
  • Provide high loading capacity and fast purifications
  • Optimized for reliable, reproducible separations
  • Compatible with simple pump-based configurations and chromatography systems such as ÄKTA design
  • Excellent choice for rapid enrichment during initial capture of proteins from start material
  • Strong Sulfopropyl (SP) cation exchanger
It is prepacked with Q Sepharose™ XL for inital capture of proteins and other biomoleculs with ion exchange (IEX) chromatography.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge