Cation exchange chromatography column, HiPrep™ SP FF
Lieferant: Cytiva
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
28-9365-44
28-9365-44EA 733 EUR InStock
28-9365-44
Cation exchange chromatography column, HiPrep™ SP FF
Chromatographiesäulen
HiPrep™ SP FF are ready to use strong cation exchange columns prepacked with SP Sepharose™ Fast Flow for fast, preparative separations of proteins and other biomolecules using ion exchange (IEX) chromatography.
  • HiPrep™ columns provide fast, reliable and reproducible preparative ion exchange separations
  • Good flow rates and high capacity
  • The industry standard for ion exchange chromatography
Compatible with single-pump configurations and chromatography systems such as ÄKTA design. Use a weak ion exchanger if the selectivity of a strong ion exchanger is insufficient.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge