Hydrophobic interaction chromatography columns, HiPrep™ Phenyl FF
Lieferant: Cytiva
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
28-9365-45
28-9365-45EA 759 EUR InStock
28-9365-45
Hydrophobic interaction chromatography columns, HiPrep™ Phenyl FF
Chromatographiesäulen
HiPrep™ Phenyl FF 16/10, are prepacked Phenyl Sepharose™ 6 Fast Flow columns ready to use for hydrophobic interaction chromatography (HIC).
  • 20 ml HIC columns provide fast, preparative separation of proteins and other biomolecules
  • Provide high capacity, excellent flow properties, and reliable, reproducible separations
  • Phenyl Sepharose™ 6 Fast Flow is a well-established, standard aromatic HIC medium for capture and intermediate purification requiring low to medium hydrophobicity
  • Two levels of substitution help find optimal selectivity and binding capacity for a given application
  • Compatible with simple pump-based configurations or chromatography system
The columns are designed for initial and intermediate purification steps, removing bulk impurities.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge