Hydrophobic interaction chromatography columns, HiPrep™ Butyl FF
Lieferant: Cytiva
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
28-9365-47
28-9365-47EA 759 EUR InStock
28-9365-47
Hydrophobic interaction chromatography columns, HiPrep™ Butyl FF
Chromatographiesäulen
HiPrep™ Butyl FF (High Sub) 16/10, are prepacked Butyl Sepharose™ 4 Fast Flow columns ready to use for hydrophobic interaction chromatography (HIC).
  • 20 ml HIC columns provide fast, preparative separation of proteins and other biomolecules
  • Provide high capacity, excellent flow properties, and reliable, reproducible separations
  • Butyl Sepharose™ 4 Fast Flow is a well-established, standard aliphatic HIC medium for capture and intermediate purification
  • Compatible with simple pump-based configurations or chromatography system such as ÄKTA design
The columns are designed for initial and intermediate purification, removing bulk impurities.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge