Precast gels, DIGE Gel
Lieferant: Cytiva
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
28-9374-51
28-9374-51EA 589 EUR InStock
28-9374-51
Precast gels, DIGE Gel
Elektrophorese-Gele Fertiggele
DIGE gel is a 12,5% precast polyacrylamide gel in a low fluorescent glass cassette for second dimension 2-D electrophoresis and specially developed for 2-D DIGE analysis. DIGE gel and DIGE buffer kit are tools to increase the quality, reproducibility, and convenience of 2-D DIGE using Ettan™ DIGE system.
  • Ready to use products with a long shelf-life that saves you time by eliminating the need to make buffers, cast gels, and wash glass plates
  • Provides high sensitivity and high resolution protein spot maps of equal quality compared to homecast polyacrylamide gels
  • Runs with DIGE buffer kit result in spot maps with higher reproducibility compared to homecast polyacrylamide gels
Lieferumfang: Please be aware that this product may be shipped 30 days before the expiry date. For more information about batch specific expiry dates please contact Avantor technical service.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge