Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Hydrophobic interaction chromatography columns, HiScreen™ Butyl HP

Lieferant: Cytiva

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

28-9782-42
28-9782-42EA 183 EUR InStock
28-9782-42
Hydrophobic interaction chromatography columns, HiScreen™ Butyl HP
Chromatographiesäulen
HiScreen Butyl HP columns are prepacked with Butyl Sepharose High Performance hydrophobic interaction chromatography (HIC) resins. The columns are an excellent choice for method optimization and parameter screening.

  • Prepacked, ready-to-use columns for convenient process development optimization
  • 10 cm bed height of HiScreen columns is designed to allow method optimization and parameter screening
  • Easily connected in series to achieve 20 cm bed height
  • Small bed volume gives fast results and minimal sample/buffer consumption
  • Reproducible results, scalable to BioProcess columns packed with the same media using the same linear fluid velocity

Phenyl Sepharose High Performance is an aromatic HIC resin, designed for intermediate and polishing step purification steps when high resolution has priority.

The small column volume of 4,7 ml and bed height of 10 cm make HiScreen columns excellent tools for method optimization, parameter screening, robustness testing, and convenient scale-up. Process fluid velocities can be applied, since the 10 cm bed height gives enough residence time and the results can then serve as basis for linear process scale-up.

If necessary, two columns can easily be connected in series to give a bed height of 20 cm.

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge