Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Ion exchange chromatography column, HiPrep™ Q HP

Lieferant: Cytiva

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

29-0181-82
29-0181-82EA 813 EUR InStock
29-0181-82
Ion exchange chromatography column, HiPrep™ Q HP
Chromatographiesäulen
HiPrep Q HP is a 20 ml strong anion exchange chromatography column for high resolution small-scale protein purification.

  • Packed with Q Sepharose High Performance strong quarternary ammonium anion exchange resin.
  • Small (34 µm) bead size delivers high-performance, high-resolution purifications.
  • 20 ml HiPrep column format compatible with single-pump configurations and chromatography systems such as ÄKTA systems

The Q functional group is a quaternary amino group coupled to the matrix via chemically stable ether linkages. Q Sepharose High Performance is a strong anion exchanger charged with high loading capacity over broad pH ranges.

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge