Ion exchange chromatography column, HiPrep™ SP HP
Lieferant: Cytiva
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
29-0181-83
29-0181-83EA 813 EUR InStock
29-0181-83
Ion exchange chromatography column, HiPrep™ SP HP
Chromatographiesäulen
HiPrep SP HP 16/10 is a strong cation exchanger designed for preparative ion exchange chromatography separations. This 20 mL column is prepacked with SP Sepharose High Performance chromatography resin. The resin is based on rigid, highly cross-linked beaded agarose with a mean bead diameter of 34 µm for high resolution separations.
  • For high-performance preparative scale separations where high efficiency and high resolution are desired
  • Highly cross-linked agarose provides excellent chemical and physical stability
  • Slim, modern design allows for easy handling and connection
High mechanical strength of the matrix allows for high flow rates, together with outstanding resolution. Capacities vary according to the protein applied and its concentration. Loading capacity will also vary depending on the salt concentration, pH, and temperature at which the protein is applied.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge