Hydrophobic interaction chromatography columns, HiTrap™ Capto™ HIC selection kit
Lieferant: Cytiva
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
29-3210-87
29-3210-87EA 733 EUR InStock
29-3210-87
Hydrophobic interaction chromatography columns, HiTrap™ Capto™ HIC selection kit
Chromatographiesäulen
The Capto hydrophobic interaction chromatography (HIC) selection kit has a set of five 1 ml ready-to-use HiTrap™ columns for resin screening and small-scale protein purification. The HiTrap™ Capto HIC selection kit is useful in process development or research applications.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge