Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Size exclusion chromatography columns, HiLoad™ 16/600 Superose™ 6 Prep Grade (pg)

Lieferant: Cytiva

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

29-3239-52
29-3239-52EA 2843 EUR InStock
29-3239-52
Size exclusion chromatography columns, HiLoad™ 16/600 Superose™ 6 Prep Grade (pg)
Chromatographiesäulen
HiLoad™ 16/600 Superose™ 6 prep grade (pg) is a prepacked size exclusion chromatography (SEC) column for high-resolution purification of large biomolecules and protein complexes. The column is an excellent choice for purification of sample volumes up to 5 ml.

  • Convenience and reproducibility: can be used with standalone pump or a chromatography system
  • High-resolution separations: purification is possible across an exceptionally broad molecular-weight range
  • Scalable: separations can be scaled up using Superose™ 6 prep grade resin
  • ÄKTA system compatible: fittings (0.062") for direct connection to ÄKTA systems and other chromatography systems

This column is packed with Superose™ 6 pg resin, which provides good resolution separations of proteins and other biomolecules according to size. The wide fractionation range of the resin enables reproducible and robust separations in the molecular weight range from 5000 up to 5000000.

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge