Annexin V conjugates
Lieferant: BIOTIUM
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
29003 29004 29001 29005 29008 29010 29011 29012 29013 29014 99902 29045-100UL 29045-500UL 29009 29057-100UL 29057-500UL 29069 29083 29001-ML
29003.EA 325 EUR InStock
29003. 29004. 29001. 29005. 29008. 29010. 29011. 29012. 29013. 29014. 99902. 29045-100UL 29045-500UL 29009. 29057-100UL 29057-500UL 29069. 29083. 29001-ML
Annexin V conjugates
Proteinkonjugate und Markierungen
Annexin V fluorescent conjugates can be used to detect apoptotic cells. Annexin V is available conjugated to a wide variety of dyes.
  • Detect apoptotic cells with fast, reliable detection of phosphatidylserine externalisation
  • Conjugates with wide choice of CF® dye colours, plus biotin, R-PE, APC, and other fluorescent dyes
  • Biotium's CF® dyes have superior brightness, photostability, and are water-soluble
  • Can be used for fluorescence microscopy, live cell imaging, and flow cytometry
Annexin V fluorescent conjugates can be used to detect apoptotic cells. The human anticoagulant Annexin V is a 35 to 36 kDa calcium-dependent phospholipid-binding protein with high affinity for phosphatidylserine (PS). In normal viable cells, PS is located on the inner leaflet of the cytoplasmic membrane. In apoptotic cells, however, PS is translocated from the inner to the outer leaflet of the plasma membrane, where it can be detected by fluorescently labelled Annexin V.
Biotium’s next-generation CF® dyes were designed to be highly water-soluble with advantages in brightness and photostability compared to Alexa Fluor®, DyLight®, and other fluorescent dyes.
Order Now
PRODUKT
TECHNISCHE DATEN
EGÄNZENDE PRODUKTE
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge