Schutzfolie, Bytac
Lieferant: Saint-Gobain Performance plastics
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
D1069324
293-4331EA 1850 EUR InStock
293-4331
Schutzfolie, Bytac
Oberflächenschutz Oberflächenschutz
Self-adhesive laminate, consisting of a chemical-resistant FEP film surface and a vinyl substrate for protecting all common work surfaces. Can be easily peeled away without leaving any adhesive residue.
Temperature resistance: Vinyl: Up to a maximum of 93 °C ; Aluminium: Up to 182 °C (at a maximum of one hour).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge