30753.36
30753.A3
30753.22
30753.36EA
54.5
EUR
InStock
30753.36
30753.A3
30753.22
Ammoniumbenzoat ≥99%
Ammoniumbenzoat
Purity: 99%
CAS No.: 1863-63-4
Molecular Formula: C6H5CO2NH4
Formula Weight: 139.15
Flash Point: 110°(230°F)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: White to off-white
Odour: Weak
Melting Point: 192-198° dec.
Merck Reference: 14,496
MDL No.: MFCD00013069