31068.AP
31068.K2
31068.M1
31068.APEA
37.7
EUR
InStock
31068.AP
31068.K2
31068.M1
1-Butanol ≥99,4% ACS
1-Butanol
Purity: 99.4+%
CAS No.: 71-36-3
Molecular Formula: CH3(CH2)3OH
Formula Weight: 74.12
Flash Point: 35°(95°F)
Physical Form: Liquid
Density: 0.810
Refractive Index: 1.3990
Odour: Alcohol-like
pH: 7
Melting Point: -90 to -89°
Boiling Point: 117-118°
Beilstein Reference: 969148
Merck Reference: 14,1540
UN No.: 1120
MDL No.: MFCD00002964