Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Phenolphthalein 0,5 wt, % in ethanol: water (1:1), Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Achtung

MQ200

319236-100ML 319236-6X100ML
319236-100MLEA 37.7 EUR InStock
319236-100ML 319236-6X100ML
Phenolphthalein 0,5 wt, % in ethanol: water (1:1), Sigma-Aldrich®
Phenolphthalein
Phenolphthalein is an acid base indicator. It is a weak acid, which is colorless in acidic solution and turns puce pink in alkaline solution. Phenolphthalein is genotoxic. It might cause oxidative damage and binds to estrogen receptors. Phenolphthalein solution is a synthetic indicator. It is colorless in acidic and neutral conditions. Phenolphthalein gives pink color when added to a base, therefore it is considered as a base indicator.

Phenolphthalein solution has been used to measure total titratable acids by titration method.


Formel: C₂₀H₁₄O₄
MW: 318.33 g/mol
Melting Pt: –35 °C
Dichte: 0.918 g/ml (25 °C)
Flash-Pt: 24 °C (geschlossener Tiegel)
Storage Temperature: Room Temperature
MDL Number: MFCD00005913
CAS-Nummer: 77-09-8
EINECS: 201-004-7
UN: 1170
ADR: 3 III
Merck Index: 14,07243
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1be6babe565245bf70948c09d6ff986d849709c0&vid=1be6babe565245bf70948c09d6ff986d849709c0&prid=web&clid=SAPDAM
Order Now
Spezifikation Testergebnisse
Appearance (Colour) Colourless
Appearance (Form) Liquid
Visual Interval
pH 8.0 Colorless
Pass
Visual Interval
pH 8.2 Pale Pink
Pass
Visual Interval
pH 8.6 Pink
Pass
Visual Interval
pH 10.0 Red
Pass

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge