VWR® AVS TITRINORM Buffer Solution, pH 9 (at 20 °C), Borate free
Lieferant: VWR Chemicals
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
32037.297EA 46.8 EUR InStock
32037.297
VWR® AVS TITRINORM Buffer Solution, pH 9 (at 20 °C), Borate free
Puffer pH-Puffer
Solution prepared from AnalaR® NORMAPUR® grade analytical reagents.
  • Borate free
  • Reference temperature 20 °C
  • True analytical value on the label to an accuracy of ±0,02 pH units
  • Traceable to SRM from NIST
  • Batch number and packing date for traceability
  • Certificate of Analysis available at website
Bestellinformation: If significantly large volumes are required please contact your local Avantor sales office for a customised offer.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge