32471.K2
32471.K7
32471.K2EA
106
EUR
InStock
32471.K2
32471.K7
o-Xylol ≥96% für die Spektrophotometrie
o-Xylol
Purity: 96% min
CAS No.: 95-47-6
Molecular Formula: C8H10
Formula Weight: 106.17
Flash Point: 32°(89°F)
Physical Form: Liquid
Density: 0.877
Refractive Index: 1.5050
Appearance: Colourless
Melting Point: -25°
Boiling Point: 143-145°
Beilstein Reference: 1815558
Merck Reference: 14,10081
UN No.: 1307
MDL No.: MFCD00008519